Would you like to log in again?
DNase Test Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M482-500G M482-500G
95021-892EA 223.26 USD InStock
95021-892 CA95021-892
DNase Test Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the detection of deoxyribonuclease activity of bacteria and fungi, and especially for identification of pathogenic staphylococci.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty