Would you like to log in again?
Christensen Citrate Sulfite Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M495-500G M495-500G
95021-904EA 181.62 USD InStock
95021-904 CA95021-904
Christensen Citrate Sulfite Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of enteric bacilli on the basis of citrate utilization and hydrogen sulfide production.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty