Would you like to log in again?
MacConkey Broth, Double Strength, with Neutral Red, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M539-500G M539S-500G M539S-500G M539-500G
95021-964EA 165.84 USD InStock
CA95021-966 95021-964 95021-966 CA95021-964
MacConkey Broth, Double Strength, with Neutral Red, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the primary isolation of coliforms from large samples such as water or waste water.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty