Would you like to log in again?
Caffeic Acid Ferric Citrate Test Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M563-500G M563-500G
95022-002EA 586.68 USD InStock
CA95022-002 95022-002
Caffeic Acid Ferric Citrate Test Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation, presumptive identification, and differentiation of Cryptococcus neoformans from other species.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty