Would you like to log in again?
FNA Medium, Dehydrated, HiMedia
Supplier: HiMedia Laboratories
M565-500G M565-500G
95022-006EA 216.01 USD InStock
CA95022-006 95022-006
FNA Medium, Dehydrated, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the differentiation of Pseudomonas from other bacilli by the ability to reduce nitrates or nitrites to nitrogen gas (denitrification) and detection of fluorescein pigment.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty