Would you like to log in again?
Phenol Red Mannitol Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
M571-5KG M571-500G M571-5KG M571-2.5KG M571-2.5KG M571-500G
89225-890EA 1291.1 USD InStock
89225-890 CA89225-888 CA95022-010 95022-010 89225-888 CA89225-890
Phenol Red Mannitol Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Phenol red mannitol agar is used for studying mannitol fermentation by the pure cultures of microorganisms.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty