Would you like to log in again?
Violet Red Bile Glucose Agar, HiMedia Laboratories
Supplier: HiMedia Laboratories
M581-5KG M581-2.5KG M581-500G M581-500G M581-5KG M581-2.5KG
89225-916EA 596.88 USD InStock
89225-916 CA89225-916 89225-918 CA95022-026 95022-026 CA89225-918
Violet Red Bile Glucose Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Violet red bile glucose agar without lactose is used for detection and enumeration of Enterobacteriaceae in raw foods and clinical samples.
  • Appearance: Light yellow to pinkish beige homogeneous free flowing powder
  • Gelling: Firm, comparable with 1.2% agar gel
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty