Would you like to log in again?
Motility Nitrate Medium, Buffered, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M630I-500G M630I-500G M630-500G M630-500G
95022-106EA 138.28 USD InStock
95022-106 CA95022-108 95022-108 CA95022-106
Motility Nitrate Medium, Buffered, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and detection of Clostridium perfringens based on motility and nitrate test.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty