Would you like to log in again?
Bennet’s Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M694-500G M694-2.5KG M694-5KG M694-5KG M694-500G M694-2.5KG
95022-168EA 285.55 USD InStock
CA95022-168 95022-168 89226-196 CA89226-198 CA89226-196 89226-198
Bennet’s Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the cultivation and enhancement of sporulation of Nocardia and Streptomyces.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty