Would you like to log in again?
Chu’s Medium No. 10 Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M697-500G M697-100G M697-500G M697-100G
61000-150EA 96.5 USD InStock
CA61000-150 61000-150 CA95022-172 95022-172
Chu’s Medium No. 10 Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For culturing blue-green algae.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty