Would you like to log in again?
Hemmes Medium Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M775-500G M775-500G
95022-238EA 406.39 USD InStock
95022-238 CA95022-238
Hemmes Medium Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the biochemical differentiation of Salmonella and Shigella species based on dextrose, lactose, sucrose fermentation, motility, hydrogen sulfide, indole, and urease production
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty