Would you like to log in again?
Blood Agar Base No.2, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M834-500G M834-500G
95022-328EA 189.42 USD InStock
CA95022-328 95022-328
Blood Agar Base No.2, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
After the addition of blood, the medium permits maximum recovery of streptococci, pneumococci, and other fastidious pathogenic microorganisms without interfering with hemolytic reactions.
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty