Would you like to log in again?
Buffered Charcoal Agar Base, Modified, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M892-500G M892-500G
95022-426EA 195.56 USD InStock
95022-426 CA95022-426
Buffered Charcoal Agar Base, Modified, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and cultivation of Legionella species from clinical and other specimens.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty