Would you like to log in again?
Malt Extract Agar Base, Modified as per Thom and Church, Dehydrated Culture Media, HiMedia
Supplier: HiMedia Laboratories
M995-500G M995-500G
95022-610EA 175.67 USD InStock
CA95022-610 95022-610
Malt Extract Agar Base, Modified as per Thom and Church, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and cultivation of yeasts and molds.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty