Would you like to log in again?
Antibiotic Assay Medium J Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
MM1666-500G MAP1666-500G
95022-740EA 224.27 USD InStock
CA95022-740 95022-740
Antibiotic Assay Medium J Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
General-purpose medium for cultivation of a wide variety of microorganisms
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Sterility testing media are general-purpose media used for cultivation of a wide variety of microorganisms. They are also used for sterility testing of molds and lower bacteria.
Also for sterility testing in pharmaceutical procedures in accordance with IP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty