Would you like to log in again?
Bismuth Sulfite Agar Medium, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
MU027-500G MU027-500G
95022-778EA 201.44 USD InStock
95022-778 CA95022-778
Bismuth Sulfite Agar Medium, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of salmonellae from feces, urine, sewage, and other materials, in accordance with USP.
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty