Would you like to log in again?
Brain Heart CC Agar, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
M209-500G M209-500G
95022-890EA 1353.01 USD InStock
CA95022-890 95022-890
Brain Heart CC Agar, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and cultivation of fastidious pathogenic fungi, such as Histoplasma capsulatum and Balstomyces dermatitidis, from specimens heavily contaminated with bacteria.
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty