Would you like to log in again?
Folic Acid Inoculum Medium, Dehydrated, HiMedia
Supplier: HiMedia Laboratories
M541-500G M541-500G
95022-926EA 363.33 USD InStock
CA95022-926 95022-926
Folic Acid Inoculum Medium, Dehydrated, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the preparation of inoculum of Enterococcus hirae ATCC 8043 used as the test organism for folic acid assay.
  • Microbiological culture media
  • 500 g poly bottles
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty