Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rose Bengal Chloramphenicol Agar, HiMedia

Supplier: HiMedia Laboratories
M640-5KG M640-2.5KG M640-500G M640-2.5KG M640-500G M640-5KG
89226-112EA 709.22 USD InStock
89226-112 CA95022-944 CA89226-114 95022-944 CA89226-112 89226-114
Rose Bengal Chloramphenicol Agar, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
Rose bengal chloramphenicol agar is used for selective isolation and enumeration of yeasts and moulds from foods and environmental materials.

  • Appearance: Light yellow to pink homogeneous free flowing powder
  • Gelling: Firm, comparable with 1.55% agar gel
  • Colour and clarity of prepared medium: Deep pink colored clear to slightly opalescent gel forms in petri plates
  • Reaction: Reaction of 3.22% w/v aqueous solution at 25 °C, pH: 7.2 ±0.2

Mycological peptone or papaic digest of soyabean meal provides essential growth nutrients. Dextrose is the fermentable carbohydrate. Chloramphenicol has inhibitory action on gram-negative bacteria. Rose bengal dye suppresses the development of bacteria and reduces the spreading of moulds, controls the size and height of mould colonies such as Rhizopus species. Rose bengal is taken up by mould and yeast colonies thereby assist in enumeration.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty