Would you like to log in again?
HiVeg™ Peptones and Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
HiVeg™
RM027-500G RM006V-10KG RM006V-500G RM002V-500G RM006V-5KG RM014V-500G RM015V-500G RM014V-2.5KG RM188V-500G RM005-500G RM020V-500G RM007-10KG RM021V-500G RM191V-500G RM194-500G RM030-2.5KG RM005V-500G RM003V-500G RM027-10KG RM022V-500G RM030V-500G RM007-25KG RM011-500G RM326V-500G RM007-10KG RM027-25KG RM006V-25KG RM191V-500G RM007-2.5KG RM668-500G RM025-500G RM006V-5KG RM027-10KG RM014-2.5KG RM030-25KG RM005-500G RM668-500G RM021V-500G RM007-500G RM001V-500G RM027-25KG RM006V-25KG RM027-5KG RM007-25KG RM027-500G RM027-2.5KG RM005V-500G RM006V-500G RM015V-500G RM007-5KG RM025-500G RM007-5KG RM188-500G RM006V-10KG RM006V-10KG RM635V-500G RM003V-500G RM013V-500G RM023V-500G RM027-5KG RM194-500G RM012V-500G RM013V-500G RM022V-500G RM188-500G RM027-2.5KG RM002V-500G RM030-500G RM023V-500G RM030V-500G RM014V-2.5KG RM030-10KG RM020V-500G RM007-500G RM030-5KG RM011-500G RM012V-500G RM188V-500G RM014V-500G RM635V-500G RM326V-500G RM001V-500G RM007-2.5KG
61001-558EA 129.88 USD InStock
CA61001-526 CA61001-528 CA61001-522 61001-558 61001-636 CA61001-568 CA61001-602 61001-516 CA89129-476 CA95000-034 61001-520 97065-322 61001-522 CA61001-570 89229-742 61001-562 CA61001-572 CA61001-636 CA89229-574 CA61001-516 61001-508 CA89229-572 CA89229-570 CA61001-632 CA61001-512 61001-548 CA95039-298 CA61001-558 95024-188 95000-058 CA97065-322 CA61001-562 CA61001-520 61001-632 61001-512 95039-298 61001-552 61001-596 61001-616 89229-574 CA61001-548 89135-072 CA61001-588 89229-570 CA61001-544 61001-536 89229-572 CA61001-508 61001-580 CA95000-058 CA95024-202 CA95024-200 61001-586 CA89132-790 CA61001-596 95024-192 CA61001-552 61001-544 61001-588 95024-190 61001-540 61001-528 89229-740 CA61001-616 61001-568 61001-602 61001-526 CA89135-072 CA61001-536 89129-478 CA95024-204 95000-034 61001-570 CA89129-478 95024-202 89229-736 CA61001-540 89132-790 89229-738 CA61001-586 61001-578 CA61001-580 61001-572
HiVeg™ Peptones and Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based peptones and media designed to work in conjunction with or to replace standard animal-based protein sources.
  • 100% Vegetable Protein Source
  • Eliminate the Risk of BSE/TSE
  • Equivalent Performance with Animal-Based Peptones
The HiVeg™ line of products consists of uniquely formulated, 100% vegetable-based peptones and media designed to work in conjunction with or to replace standard animal-based protein sources. The products effectively eliminate BSE/TSE contamination concerns while providing results on par with animal-based products. The wide selection of peptones and media can meet the requirements of nearly any microbiology application.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty