Would you like to log in again?
Serological Pipette Rack, IBI Scientific
Supplier: IBI Scientific
PR-42 PR-40
82031-544EA 253.01 USD InStock
82031-544 95025-246
Serological Pipette Rack, IBI Scientific
Pipette Stands
These stands provide storage space for serological pipettes
Constructed of heavy-duty 6mm (1/4") clear-cast acrylic with orange shelves. Angled shelves are slightly indented for better access. Solid bottom and rubber feet for stability.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty