Would you like to log in again?
Elite Mini PAP Pen, Diagnostic BioSystems
Supplier: Diagnostic BioSystems
K042 K042
95025-252EA 137.4 USD InStock
95025-252 CA95025-252
Elite Mini PAP Pen, Diagnostic BioSystems
Stains Microscopy and staining reagents
Elite mini PAP pen is a special marking pen that provides a thin film-like green-tinged hydrophobic barrier when a circle is drawn around a specimen on a slide. This water repellent barrier keeps staining reagents localized on the tissue sections, prevents mixing of reagents when differentially staining two sections on the same slide, and allows use of less reagents per section.
  • Insoluble in alcohol and acetone
  • Finer tip than the regular PAP pen
PAP Pen markings are insoluble in alcohol and acetone. It can be optionally removed with xylene after the staining procedure is complete. The pen is especially useful when working with multiple sections on a single slide because of its finer tip.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty