Would you like to log in again?
HyClone™ SFMTransfx-293 Serum-free Media, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
HyClone™
SH30861.01 SH30860.02 SH30860.02 SH30861.01
95026-636EA 126.9 USD InStock
CA95026-636 95026-636 CA76462-538 76462-538
HyClone™ SFMTransfx-293 Serum-free Media, HyClone products (Cytiva)
Cell Culture Media
HyClone SFMTransfx-293 is a serum-free, animal derived component-free medium designed to support the growth of HEK 293 cultures and promote transfection using lipofection or similar methods. Manufactured according to cGMP guidelines.
  • Supports high transfection efficiency
  • Metabolically designed for high cell yield and recombinant protein/vector production
  • Allows for direct or sequential adaptation
HyClone SFMTransfx-293 Media is a regulatory-friendly medium developed through the Metabolic Pathway Design to support high transfection efficiency, productivity and cell density in suspension cultures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty