Would you like to log in again?
Renewables™ Sharps Disposal Containers, Covidien
Supplier: Covidien
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Renewables™
8537Y 8900MW 8507MW 8967MW
95026-678CS 211.74 USD InStock
95026-678 95026-676 15704-246 95026-670
Renewables™ Sharps Disposal Containers, Covidien
Bins Sharps Containers
Containers are manufactured using recycled plastics reclaimed from the manufacturing process
Containers meet or exceed the OSHA Needlestick Safety and Prevention Legislation, NIOSH/CDC guidelines, DOT requirements, and JCAHO standards.
These renewables™ sharps disposal containers are made of plastic.
Translucent containers allow for visual monitoring. Always-open lids allow quick and easy disposal, especially for larger syringes. Horizontal drop lids maximize use of container volume. All containers are autoclavable and include a biohazard label. Containers lock for final disposal.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty