Would you like to log in again?
illustra™ GFX™ PCR DNA and Gel Band Purification Kits
Supplier: Cytiva
illustra™ GFX™
VN33081 28-9034-70 28-9034-71 29489379 28903466 28-9034-71 28-9034-70
95026-730EA 789.34 USD InStock
MPCA29489379 CA95026-730L 95026-730 101725-244 CA95026-728L CA101725-244 95026-728
illustra™ GFX™ PCR DNA and Gel Band Purification Kits
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The illustra GFX PCR DNA and Gel Band Purification kits are for the isolation and concentration of DNA fragments from PCR mixtures, DNA-containing agarose gel bands, enzyme-based DNA modifications, and restriction enzyme digests.
  • Fast and easy-to-use method with less than 10 min hands-on time
  • Flexible 10 to 50 μl elution volume for different DNA concentration needs
  • Improved column design with increased maximum volume
  • Holding of up to 800 μl liquid volume (950 μl without a cap)
  • Two choices of elution buffer provided to fit downstream applications
  • Color-coded caps for ease of use
illustra GFX PCR DNA and Gel band purification kit is designed for the rapid purification and concentration of PCR products or DNA fragments ranging in size from 50 bp to 10 kb. This kit can be used to purify DNA from reaction volumes up to 100 μl or agarose gel slices up to 900 mg. The kit combines a versatile chaotropic buffer with a glass fiber matrix supported in a spin column for the purification of DNA from both solution and agarose gel. Typical recoveries range from 60 to 80% for DNA fragments from agarose gel to as high as 95% for PCR products from solution. DNA purity is exceptional, 99.5% of contaminants are removed.
Highly pure DNA is ready for direct use in sequencing, PCR, labeling, restriction enzyme digestion, and cloning. Addition of a visual color indicator to the capture buffer ensures optimal pH for maximum DNA binding and recovery.
Ordering information: Kit contains GFX columns (2×50), collection tubes (2×50). color-coded bottles of capture buffer (blue, 55 ml), wash buffer (yellow, 25 ml), and two elution buffers each 12 ml (Tris-HCl and sterile water) and an instruction booklet.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty