Would you like to log in again?
Buffered Peptone Water, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia Laboratories
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
M614-500G M614-5KG M614S-500G M614-2.5KG M614-500G M614-10KG M614-25KG M614-10KG M614S-500G M614-25KG M614-2.5KG
95026-848EA 790 USD InStock
CA95059-662 95026-848 95059-662 CA95022-078 CA95022-076 95022-078 CA71002-740 95022-076 CA89130-344 71002-740 89130-344
Buffered Peptone Water, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
A pre-enrichment medium for increasing the recovery of injured Salmonella species from foods prior to selective enrichment and isolation.
  • Microbiological culture media
  • Packed in poly bottles
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty