Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
AFB Staining Kit for histology
Supplier: Diagnostic Biosystems
KT001 KT001
95027-170EA 397.46 USD InStock
95027-170 CA95027-170
AFB Staining Kit for histology
AFB Staining Kit
This kit stains acid-fast bacteria and tubercle bacilli of Mycobacterium genus in paraffin tissue sections
Tubercle bacilli and other acid-fast bacteria stain bright red and the background stains light green.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty