Would you like to log in again?
Fite Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic BioSystems
KT013 KT013
95027-186EA 434.72 USD InStock
CA95027-186 95027-186
Fite Stain Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains Lepra bacillus in paraffin tissue sections
The Lepra bacillus stains red, while the background stains blue.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty