Would you like to log in again?
Fontana Masson Kit, Diagnostic Biosystems
Supplier: Diagnostic BioSystems
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KT014 KT014
95027-188EA 509.65 USD InStock
95027-188 CA95027-188
Fontana Masson Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains argentaffin cells and melanin in paraffin tissue sections
The argentaffin cells and melanin granules stain brown to black, while the nuclei stain pink.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty