Would you like to log in again?
GMS Kit, Diagnostic Biosystems
Supplier: Diagnostic BioSystems
KT015 KT015
95027-190EA 541.25 USD InStock
95027-190 CA95027-190
GMS Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains fungi and Pneumocystis carinii in paraffin tissue sections
The fungi and Pneumocystis carinii stain gray to black, while the background stains green.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty