Would you like to log in again?
Oil Red O Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic BioSystems
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
KT025 KT025
95027-208EA 493.73 USD InStock
95027-208 CA95027-208
Oil Red O Histology Stain Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains fat cells and neutral fat in frozen tissue sections
The fat cells and neutral fat stains red, while the nuclei stain blue.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty