Would you like to log in again?
Pneumocystis Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic BioSystems
KT030 KT030
95027-216EA 406.15 USD InStock
CA95027-216 95027-216
Pneumocystis Histology Stain Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains Pneumocystis carinii in cytology smears as well as paraffin tissue sections
The Pneumocystis carinii stains violet to purple and the connective tissue stains blue to green.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty