Would you like to log in again?
CytoScan™ WST-1 Cell Proliferation Assay, G-Biosciences
Supplier: G-Biosciences
786-212 786-212
95029-188EA 300.5 USD InStock
CA95029-188 95029-188
CytoScan™ WST-1 Cell Proliferation Assay, G-Biosciences
Assays Cellular Assays
G-Biosciences' CytoScan™ WST-1 Cell Proliferation Assay is a sensitive and accurate assay for cell proliferation and cytotoxicity
G-Biosciences' CytoScan™ WST-1 Cell Proliferation Assay principle is based upon the reduction of the high sensitivity tetrazolium salt, WST-1, to formazan by cellular dehydrogenases. The generation of the dark yellow colored formazan is measured at 420 to 480nm (optimal at 440nm) and is directly correlated to cell number.
G-Biosciences' CytoScan™ WST-1 Cell Proliferation Assay is supplied in a kit that contains components sufficient for performing up to 500 assays.
The assay is highly convenient as it is performed in a single cell tissue culture well and requires no washing, harvesting or solubilization of cells. Adherent or suspension cells are cultured in a microplate and then incubated with WST-1 and the assay is monitored with a spectrophotometer.
Ordering information: Accessories available
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty