Would you like to log in again?
PhosphataseArrest™ Phosphatase Inhibitor Cocktails, G-Biosciences
Supplier: G-Biosciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
786-450 786-451 786-452
95029-238EA 362.68 USD InStock
CA95029-242 95029-238 CA95029-238 95029-240 95029-242 CA95029-240
PhosphataseArrest™ Phosphatase Inhibitor Cocktails, G-Biosciences
Cell Culture Supplements and Additives
G-Biosciences' PhosphataseArrest™ phosphatase inhibitor cocktails are ready-to-use, 100X concentrated solutions that are simply added to your extraction buffers or samples
G-Biosciences' PhosphataseArrest™ I is a broad spectrum phosphatase inhibitor cocktail consisting of five phosphatase inhibitors that target serine/threonine specific, tyrosine specific and dual specificity phosphatases. PhosphataseArrest™ I is a stablized solution of sodium fluoride, sodium orthovanadate, sodium pyrophosphate, β-glycerophosphate & sodium molybdate.
G-Biosciences' PhosphataseArrest™ II is a phosphatase inhibitor cocktail consisting of five phosphatase inhibitors that target acid, alkaline and tyrosine phosphatases. PhosphataseArrest™ II contains optimized concentrations of sodium fluoride, sodium tartrate, sodium orthovanadate, imidazole & sodium molybdate.
G-Biosciences' PhosphataseArrest™ III is a phosphatase inhibitor cocktail consisting of three phosphatase inhibitors that target alkaline and serine/threonine phosphatases. PhosphataseArrest™ III is a stable, convenient solution of cantharidin, p-bromotetramisole oxalate and calyculin.
These phosphatase inhibitor cocktails are compatible with most phosphatase assays, and there is no resuspension required.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty