Would you like to log in again?
Phosphatase Assay Kit, G-Biosciences
Supplier: G-Biosciences
786-453
95029-244EA 533.35 USD InStock
95029-244 CA95029-244
Phosphatase Assay Kit, G-Biosciences
Assays Enzyme Assays
G-Biosciences' Phosphatase Assay kit is designed to measure the activity of phosphatases in biological samples and to screen for agonists and inhibitors of phosphatases.
G-Biosciences' Phosphatase Assay kit uses para-nitrophenyl phosphate (pNPP), a chromogenic substrate for most phosphatases, including alkaline phosphatases, acid phosphatases, protein tyrosine phosphatases and serine/threonine phosphatases. The phosphatases remove the phosphate group to generate p-nitrophenol, which is deprotonated under alkaline conditions in order to produce p-nitrophenolate, which has strong absorption at 405nm.
The supplied kit components are sufficient for performing up to 1000 assays in 96-well plate format, and are easily adaptable to cuvettes or 384-well plates.
Ordering information: Accessories available
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty