Would you like to log in again?
Human Single Tissue Single Species Blot, Tumor Tissue, G-Biosciences
Supplier: G-Biosciences
TB72 TB86 TB72 TB87 TB88 TB82 TB74 TB84 TB85 TB90 TB84 TB80 TB75 TB89 TB76 TB54 TB77 TB88 TB89 TB74 TB78 TB91 TB76 TB87 TB85 TB73 TB54 TB78 TB77 TB91 TB73 TB90 TB86 TB75 TB80 TB81 TB83 TB81 TB83 TB82
95029-390EA 517.78 USD InStock
95029-390 95029-392 95029-370 CA95029-380 95029-394 95029-372 CA95029-382 CA95029-362 95029-396 95029-374 CA95029-384 95029-366 95029-388 CA95029-396 CA95029-374 95029-400 95029-368 CA95029-398 82023-238 CA95029-378 95029-380 CA95029-390 95029-382 95029-362 CA95029-370 CA95029-392 95029-384 95029-386 CA95029-394 CA95029-372 95029-364 95029-398 CA95029-386 CA95029-364 95029-378 CA95029-366 CA95029-388 CA95029-368 CA95029-400 CA82023-238
Human Single Tissue Single Species Blot, Tumor Tissue, G-Biosciences
Blotting Kits
G-Biosciences offers a selection of ready-to-screen, human, single tissue blots, from tumor tissues.
  • Blots are ready to be blocked and probed with the antibodies of choice
  • Each blot contains two protein lanes, a protein marker lane and a lane with GenLysate™ sample
G-Biosciences premade western blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty