Would you like to log in again?
Human Single Tissue Single Species Blot, Normal and Tumor Tissue, G-Biosciences
Supplier: G-Biosciences
TB93 TB100 TB98 TB96 TB94 TB95 TB97 TB94 TB95 TB98 TB97 TB96 TB93 TB92 TB100 TB92
95029-410EA 895.09 USD InStock
95029-410 CA95029-410 95029-412 CA95029-412 95029-402 95029-414 CA95029-414 95029-404 CA95029-402 95029-406 CA95029-404 95029-418 CA95029-418 95029-408 CA95029-406 CA95029-408
Human Single Tissue Single Species Blot, Normal and Tumor Tissue, G-Biosciences
Blotting Kits
G-Biosciences offers a selection of ready-to-screen, human, single tissue blots, from both normal and tumor tissues.
  • Blots are ready to be blocked and probed with the antibodies of choice
  • Each blot contains two protein lanes, a protein marker lane and a lane with GenLysate™ sample
G-Biosciences premade western blots are prepared with the appropriate GenLysate™ and 50 μg protein is loaded onto a 4-20% denaturing polyacrylamide gel along with a prestained molecular weight marker. After the proteins are resolved, they are transferred to a PVDF membrane, using our Efficient Western Transfer Buffer.
Ordering information: Accessories available.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty