Would you like to log in again?
GroPro PLUS™ Growth Promotion Test Suspensions, NAMSA®
Supplier: Namsa
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GroPro PLUS™
GP-10 GP25-08 GP-04 GP-06 GP25-09 GP25-03 GP25-11 GP25-07 GP-02 GP-01
19872-050EA 312.63 USD InStock
19872-050 95029-730 19872-054 95029-722 95029-724 19872-058 95029-714 95029-726 95029-706 19872-048
GroPro PLUS™ Growth Promotion Test Suspensions, NAMSA®
Microbiology Test Systems Endotoxin Tests
These ready-to-use microbial suspensions meet requirements for media growth promotion performance testing
Applications include validation tests in bacteriostasis/fungistasis, quality control challenges of media for sterility testing, challenges of media for antimicrobial effectiveness testing, and for the testing of environmental monitoring media. Intended for use in non-clinical testing only.
All suspensions comply with requirements for a population of <100 colony-forming units (CFU) per 0.1mL and are guaranteed to be within five passages of an original stock culture. Suspensions are easy to use, store, and dispose of. They do not require reconstitution or dilution.
Ordering information: Suspensions are provided as purified organisms in vials containing 25 or 100 doses each. Must be stored under refrigerated conditions from 2 to 8°C (35.6 to 46.4°F).
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty