Would you like to log in again?
Glucose Tolerance Drinks, Azer Scientific
Supplier: Azer Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
10-FP-100 10-O-100 10-FP-050 10-O-050 10-FP-075 10-SP-100 10-LL-075 10-LL-100 10-LL-050 10-O-075-CA 10-SP-050 10-O-050-CA 10-O-075
95030-170CS 348.5 USD InStock
95030-170 95030-182 CA11006-155 95030-180 CA11006-153 95030-178 95030-168 95030-176 95030-166 95030-174 95030-172 89129-898 95030-184
Glucose Tolerance Drinks, Azer Scientific
Beverages
FDA-approved, non-carbonated beverages are designed for use in conjunction with glucose tolerance tests.
  • Shatterproof Plastic Bottles
  • Graduated Labels on Bottles
  • Non-Carbonated Beverages
  • Citric Acid Free
  • Dairy and Gluten-Free
All beverages are supplied in shatterproof plastic bottles.
Beverages are gluten- and dairy-free, certified Kosher, and are manufactured according to WHO and ADA standards. Simply Pure unflavored beverage is also citric acid free. The integrity of each batch is certified by an FDA recognized independent testing laboratory. All bottles are marked with an expiration date determined via accelerated and real time stability studies.
A graduated label on each bottle indicates the amount dispensed.
Certifications: Beverages are gluten and dairy free, certified Kosher, and are manufactured according to WHO and ADA standards. Simply Pure unflavored beverage is also citric acid free. The integrity of each batch is certified by a FDA-recognized independent testing laboratory. All bottles are marked with an expiration date determined via accelerated and real-time stability studies.
Ordering information: Beverages are packaged in four shrink-wrapped six-packs per case. Beverages are packaged in four shrink-wrapped six-packs per case.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty