These cassettes are made of a high-density acetal polymer and keep specimens safely submerged in liquid
The cassettes are totally resistant to the chemical action of histological solvents. The flow-through openings allow for maximum fluid exchange and ensure proper drainage. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassettes. The anterior writing area is at a 45° angle. All cassettes colors are non-cytotoxic and non-metallic. Dimensions: 42.2Lx28.5Wx6Hmm (121/32x11/8x1/4").
Ordering information: Each case contains 10 sleeves of 75 bases and 10 bags of 75 lids. Cassette bases are packaged in transparent sleeves for quick loading into automatic labeling instruments.