Would you like to log in again?
AccuTarget™ Control siRNA Set, Bioneer
Supplier: Bioneer
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SP-1012 SP-2013 SN-1023 SS-1013 SP-2011 SS-1001 SN-1011 SP-1001 SN-1022 SN-1001 SS-1011 SP-1013 SP-2001 SP-3013 SS-1012 SP-1011 SP-3001 SP-3003 SP-3012 SP-2012 SP-2003 SS-1003 SN-1012 SN-1002 SS-1002 SP-2002 SN-1021 SN-1013 SP-1003 SN-1003 SP-3011 SP-1002 SP-3002
95030-542EA 148.88 USD InStock
95030-542 95030-586 95030-564 95030-584 95030-540 95030-562 95030-560 95030-582 95030-580 95030-528 95030-548 95030-526 95030-546 95030-524 95030-568 95030-588 95030-566 95030-544 95030-574 95030-530 95030-552 95030-572 95030-550 95030-570 95030-538 95030-536 95030-558 95030-578 95030-534 95030-556 95030-532 95030-576 95030-554
AccuTarget™ Control siRNA Set, Bioneer
Nucleic Acid Reagents RNAi
AccuTarget™ Control siRNAs are a wide range of positive control siRNAs, negative control siRNAs and control siRNA sets
Control siRNA can be fluorescently labeled for monitoring transfection efficiency.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty