Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lactic acid 85% ≥85% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: 2-Hydroxypropionic acid2-Hydroxypropanoic acidDL-Lactic acid

L1009-4CS4 L1009-4LTPL L1009-500MLP L1009-500CS6 L1009-20LT L1009-200LTBL L1009-125ML
95031-440EA 876.18 USD InStock
95031-440 75811-458 95031-506 95031-508 95031-438 95031-504 75811-460
Lactic acid 85% ≥85% FCC
Lactic acid
Lactic Acid, 85 Percent, FCC is used as a food preservative, curing agent, and flavoring agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

GHS hazard : Corrosion
UN3265


Formula: H₃CCH(OH)COOH
MW: 90.08 g/mol
Density: 1.209
Storage Temperature: Ambient
MDL Number: MFCD00004520
CAS Number: 50-21-5
UN: 3265
ADR: 8,III
Order Now
Specification Test Results
Assay (C₃H₆O₃) 81 - 89%
Chloride (Cl) max. 0.1%
Citric, Oxalic, Phosphoric and Tartaric Acid To Pass Test
Cyanide max. 5 mg/kg
Iron (Fe) max. 10 mg/kg

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty