Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 10% (w/v) NF

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Muriatic acidHydrochloric acid solution

HY105-2.5CS4 HY105-2.5LTP HY105-500CS6 HY105-200LTBL HY105-20LTBL HY105-500MLG
95031-484CS 809.88 USD InStock
95031-484 75810-490 95031-530 95031-486 95031-528 75810-488
Hydrochloric acid 10% (w/v) NF
Hydrochloric acid
Diluted Hydrochloric Acid, 10 Percent (w/v), NF is a pH acidifier excipient. 

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 


Formula: HCl
MW: 36.46 g/mol
Density: 1.05
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay (HCl, W/V) 9.5 - 10.5 %
Residue On Ignition To Pass Test
Sulfate To Pass Test
Sulfite (SO₃) To Pass Test
Heavy Metals max. 5 µg/g

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty