Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% NF

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning
Synonyms: Chlorbutanol hemihydrate

CH122-12KGBL CH122-500GM CH122-25GM CH122-50KGBL CH122-125GM CH122-2.5KG
75810-934EA 2662 USD InStock
75810-934 95032-518 700003-054 95032-522 95032-520 89078-728
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98.0% NF
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 


Formula: C₄H₇Cl₃O·0,5H₂O
MW: 186.46 g/mol
Boiling Pt: 167 °C (1013 hPa)
Melting Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00004461
CAS Number: 6001-64-5
EINECS: 200-317-6
Order Now
Specification Test Results
Assay (C₄H₇Ocl₃) 98.0 - 100.5 %
Reaction (To Litmus) Neutral
Water max. 6.0 %
Chloride (Cl) max. 0.07 %
Identification To Pass Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty