Synonyms:
Sodium Citrate, Dihydrate
, Citric acid trisodium salt dihydrate
, Sodium Citrate Dihydrate
, Sodium citrate tribasic dihydrate
, Trisodium citrate
, Sodium citrate dihydrate
, Citric acid tridosium salt dihydrate
SO165-50KGBL
SO165-12KG
SO165-500CS6
SO165-2.5KG
SO165-12KG
SO165-500GM
SO165-2.5CS4
95035-798CS
1708.5
USD
InStock
95035-798
75812-436
95035-804
89050-470
95035-800
CA89050-470
95035-802
tri-Sodium citrate dihydrate ≥99.0%, granulated USP
tri-Sodium citrate dihydrate
Sodium Citrate, Dihydrate, Granular, USP can be used as an alkalizing agent, buffering agent, emulsifier, or sequestering agent.
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.