Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 37% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms: Muriatic acidHydrochloric acid solution

HY106-4LT HY106-2.5LTG HY106-4LT-CS4 HY106-20LT HY106-2.5CS4 HY106-200LTBL HY106-500MLGL HY106-500ML-CS
95036-672CS 596.7 USD InStock
95036-672 75811-390 75811-388 95036-674 75811-384 75811-386 75811-392 75811-382
Hydrochloric acid 37% FCC
Hydrochloric acid
Hydrochloric Acid, 37 Percent, FCC is used in food processing, or as a food additive to adjust the pH. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

GHS hazard : Corrosion;Exclamation mark
UN1789


Formula: HCl
MW: 36.46 g/mol
Density: 1.200
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 36.0 - 38.0 %
Color To Pass Test
Specific Gravity To Pass Test
Iron (Fe) max. 5 mg/kg
Lead (Pb) max. 1 mg/kg

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty