Would you like to log in again?
Sed-Chek® 2 Sedrate Controls, Polymedco
Supplier: Polymedco
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sed-Chek®
ESR-2CT ESR-2CTA 261091-BDL SJ1215PLUSBDL S-1090 ESR-2CTN ESR-2SSN ESR-5CT ESR-5CTN ESR-2SSA ESR-5CTA
95037-990EA 442.6 USD InStock
95037-990 95038-106 95037-992 95037-980 95037-994 95037-982 95037-984 95037-986 75815-660 95037-988 75815-694
Sed-Chek® 2 Sedrate Controls, Polymedco
Control reagents
Sed-Chek 2 is a whole blood reference control material designed to monitor ESR procedures.
  • Helps to monitor technique as well as environmental, physical and mechanical factors
  • Improved laboratory confidence in accurate results
The erythrocyte sedimentation rate test is a variable test where the result can be affected by environmental and procedural factors. Sed-Chek 2, a bi-level whole blood control, was designed for use with manual methods and automated ESR analyzers to monitor ESR procedures. Sed-Chek 2 employs a unique cell stabilizing process which mimics the rouleaux formation of patient samples. Use of the Sed-Chek 2 ESR control will monitor technique as well as the environmental, physical and mechanical factors such as room temperature, tube position, vibration or instrument problems which adversely affect ESR results.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty