Would you like to log in again?
dNTP solutions, illustra™, Cytiva
Supplier: Cytiva
illustra™
28-4065-41 28-4065-22 28-4065-01 28-4065-32 28-4065-32 28-4065-41 28-4065-03 28-4065-01 28-4065-42 28-4065-42 28-4065-02 28-4065-12 28-4065-02 28-4065-22 28-4065-12
95038-234EA 224.36 USD InStock
CA95038-226L CA95038-252 CA95038-246L CA95038-234L 95038-234 95038-230 95038-252 95038-240 95038-250 95038-228 95038-226 95038-246 CA95038-240L CA95038-250L CA95038-228
dNTP solutions, illustra™, Cytiva
Nucleic Acid Reagents Nucleotides
illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty