Would you like to log in again?
dNTP sets, illustra™, Cytiva
Supplier: Cytiva
illustra™
28-4065-53 28-4065-52 28-4065-53 28-4065-51 28-4065-58 28-4065-57 28-4065-51 28-4065-52 28-4065-58 28-4065-57
95038-258EA 3347.15 USD InStock
CA95038-254L CA95038-258L 95038-258 CA95038-256L 95038-256 95038-254 CA95038-262L 95038-262 CA95038-260L 95038-260
dNTP sets, illustra™, Cytiva
Nucleic Acid Reagents Nucleotides
High purity dNTPs for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for consistent, high performance
  • Buffer-free, ready to use solutions at a variety of concentrations
  • Functionally tested to produce a 20,7 kb PCR amplification product from lambda DNA
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty