Would you like to log in again?
illustra™ PCR nucleotide mix dNTP set, Cytiva
Supplier: Cytiva
illustra™
28-4065-64 28-4065-60 28-4065-64 28-4065-60
95038-268EA 125.76 USD InStock
CA95038-264L CA95038-268L 95038-268 95038-264
illustra™ PCR nucleotide mix dNTP set, Cytiva
Nucleic Acid Reagents Nucleotides
PCR nucleotide mix dNTP set (2 mM, 10 mM, or 25 mM each A,C,G,T) for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready to use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty